Tisk…

Cesium orthovanadate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

39211.09 39211.18
39211.09EA 4470 CZK
39211.09 39211.18
Cesium orthovanadate ≥99.9% (metals basis)
Cesium orthovanadate

Purity: 99.9% CAS No.: 34283-69-7 Molecular Formula: Cs3VO4 Formula Weight: 513.65 Physical Form: -100 Mesh Powder UN No.: 3288 MDL No.: MFCD00210609

Vzorec: Cs₃O₄V
Storage Temperature: Ambient
MDL Number: MFCD00210609
CAS číslo: 34283-69-7
EINECS: 251-916-4
UN: 3288
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR