Tisk…

Potassium metavanadate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

39501.14 39501.22
39501.14EA 2810 CZK
39501.14 39501.22
Potassium metavanadate ≥99.9% (metals basis)
Potassium metavanadate

Purity: 99.9% CAS No.: 13769-43-2 Molecular Formula: KVO3 Formula Weight: 138.04 Physical Form: -200 Mesh Powder Odour: Odourless UN No.: 2864 MDL No.: MFCD00050038

Vzorec: KO₃V
MW: 138.04
Bod tání: 520 °C
Hustota: 2.84 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00050038
CAS číslo: 13769-43-2
EINECS: 237-388-8
UN: 2864
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR