Tisk…

Potassium hexafluorotitanate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10915.36 A10915.22 A10915.0E
A10915.36EA 1980 CZK
A10915.36 A10915.22 A10915.0E
Potassium hexafluorotitanate ≥97%
Potassium hexafluorotitanate

Purity: 97% CAS No.: 16919-27-0 Molecular Formula: F6K2Ti Formula Weight: 240.07 Boiling Point: 235-237° UN No.: 3288 MDL No.: MFCD00011380

Vzorec: F₆K₂Ti
Bod varu: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00011380
CAS číslo: 16919-27-0
EINECS: 240-969-9
UN: 3288
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR