Tisk…

Potassium hexafluorosilicate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

89023.A1 89023.A7
89023.A1EA 3090 CZK
89023.A1 89023.A7
Potassium hexafluorosilicate ≥98%
Potassium hexafluorosilicate

Purity: 98% CAS No.: 16871-90-2 Molecular Formula: K2SiF6 Formula Weight: 220.28 Physical Form: Powder Density: 2.27 pH: 3.4 Melting Point: dec. Solubility: Slightly soluble in cold water. Hydrolyzes in hot water. Insoluble in alcohol Merck Reference: 14,7638 UN No.: 2655 MDL No.: MFCD00040666

Vzorec: F₆K₂Si
Bod tání: Dec.
Hustota: 2,27
Storage Temperature: Ambient
MDL Number: MFCD00040666
CAS číslo: 16871-90-2
EINECS: 240-896-2
UN: 2655
ADR: 6.1,III
Merck Index: 14,7638

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR