Tisk…

, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

M-Clarity™ quality level = MQ200

P2222-100G P2222-1KG P2222-500G
SIALP2222-100GEA 1580 CZK
SIALP2222-100G SIALP2222-1KG SIALP2222-500G
, Sigma-Aldrich®
Hydrogenfosforečnan didraselný
Vzorec: K₂HPO₄
MW: 174.18
Bod tání: 340
Hustota: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS číslo: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR