Tisk…

Molybdenan draselný ≥95%

Dodavatel: Thermo Fisher Scientific
Varování

48112.22 48112.36
48112.22EA 2250 CZK
48112.22 48112.36
Molybdenan draselný ≥95%
Molybdenan draselný

Purity: 95% CAS No.: 13446-49-6 Molecular Formula: K2MoO4 Formula Weight: 238.14 Physical Form: Powder Density: 2.3 Melting Point: 919° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,7647 MDL No.: MFCD00011370

Vzorec: K₂MoO₄
Bod tání: 919 °C
Hustota: 2,3
Storage Temperature: Ambient
MDL Number: MFCD00011370
CAS číslo: 13446-49-6
EINECS: 236-599-2
Merck Index: 14,7647

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR