Hydrogendifluorid draselný ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Potassium hydrogen fluoride, Potassium bifluoride
Purity: 99+%
CAS No.: 7789-29-9
Molecular Formula: KHF2
Formula Weight: 78.10
Physical Form: Crystalline
Density: 2.370
pH: 1
Melting Point: 239° dec.
Solubility: Soluble in water (39g/100ml), dilute alcohol. Insoluble in absolute alcohol
Merck Reference: 14,7610
UN No.: 1811
MDL No.: MFCD00011403
Electrolyte, flux, catalyst in alkylation of benzene with olefins, and in the synthesis of pure KF
Vzorec:
HF₂K Hustota: 2,370 Storage Temperature: Ambient |
MDL Number:
MFCD00011403 CAS číslo: 7789-29-9 EINECS: 232-156-2 UN: 1811 ADR: 8,II Merck Index: 14,7610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...