10-Methylphenothiazine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B21648.06
B21648.14
B21648.06EA
2130
CZK
B21648.06
B21648.14
10-Methylphenothiazine ≥98%
10-Methylphenothiazine
Purity: 98%
CAS No.: 1207-72-3
Molecular Formula: C13H11NS
Formula Weight: 213.30
Melting Point: 100-103°
MDL No.: MFCD00041836
Vzorec:
C₁₃H₁₁NS MW: 213,3 g/mol Bod varu: 342 °C (1013 hPa) Bod tání: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041836 CAS číslo: 1207-72-3 EINECS: 214-896-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...