Phenothiazine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12517.0I
A12517.30
A12517.36
A12517.0IEA
10990
CZK
A12517.0I
A12517.30
A12517.36
Phenothiazine ≥98%
Phenothiazine
Purity: 98+%
CAS No.: 92-84-2
Molecular Formula: C12H9NS
Formula Weight: 199.28
Melting Point: 182-186°
Boiling Point: 235°/21mm
Merck Reference: 14,7252
MDL No.: MFCD00005015
Vzorec:
C₁₂H₉NS Bod varu: 235 °C (21 mmHg) Bod tání: 182…186 °C Hustota: 1,34 g/cm³ (20 °C) Bod vzplanutí: 202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005015 CAS číslo: 92-84-2 EINECS: 202-196-5 Merck Index: 14,7252 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...