Tisk…

4-(Dimethylamino)antipyrine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,3-Dimethyl-4-(dimethylamino)-1-phenyl-3-pyrazolin-5-one, 4-(Dimethylamino)phenazone

B21436.0B B21436.18 B21436.30
B21436.0BEA 24470 CZK
B21436.0B B21436.18 B21436.30
4-(Dimethylamino)antipyrine ≥98%
4-(Dimethylamino)antipyrine

Purity: 98+% CAS No.: 58-15-1 Molecular Formula: C13H17N3O Formula Weight: 231.30 Odour: Odourless Melting Point: 105-109° UN No.: 2811 MDL No.: MFCD00003142

Vzorec: C₁₃H₁₇N₃O
MW: 231.3
Bod tání: 105…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003142
CAS číslo: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1 III
Merck Index: 12,00495

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR