Tisk…

PIPES disodium salt, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

P3768-100G P3768-1KG P3768-250G P3768-25G P3768-500G
SIALP3768-100GEA 9670 CZK
SIALP3768-100G SIALP3768-1KG SIALP3768-250G SIALP3768-25G SIALP3768-500G
PIPES disodium salt, Sigma-Aldrich®
PIPES disodium salt
Vzorec: C₈H₁₆N₂Na₂O₆S₂
MDL Number: MFCD00064349
CAS číslo: 76836-02-7
EINECS: 278-562-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR