Tisk…

1-(3-Hydroxypropyl)piperazine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Piperazinepropanol, 3-Piperazin-1-ylpropanol, 3-(Piperazin-1-yl)propan-1-ol

H50290.03 H50290.06
H50290.03EA 2260 CZK
H50290.03 H50290.06
1-(3-Hydroxypropyl)piperazine ≥98%
1-(3-Hydroxypropyl)piperazine

Purity: 98% CAS No.: 5317-32-8 Molecular Formula: C7H16N2O Formula Weight: 144.22 Melting Point: 48-52° MDL No.: MFCD00023132

Vzorec: C₇H₁₆N₂O
MW: 144,22 g/mol
Bod varu: 264 °C (1013 hPa)
Bod tání: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00023132
CAS číslo: 5317-32-8
EINECS: 226-176-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR