Tisk…

Homopiperazine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4-Diazepane, 1,4-Diazacycloheptane, Perhydro-1,4-diazepine

M-Clarity™ quality level = MQ200

H16604-100G H16604-25G
SIALH16604-100GEA 5320 CZK
SIALH16604-100G SIALH16604-25G
Homopiperazine, Sigma-Aldrich®
Homopiperazine
Vzorec: C₅H₁₂N₂
MW: 100,16 g/mol
Bod varu: 169 °C (1013 hPa)
Bod tání: 43 °C
MDL Number: MFCD00006933
CAS číslo: 505-66-8
EINECS: 208-016-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR