Cyclopropyl methyl ketone ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Cyclopropylethanone
A13540.14
A13540.22
A13540.36
A13540.14EA
1090
CZK
A13540.14
A13540.22
A13540.36
Cyclopropyl methyl ketone ≥99%
Cyclopropyl methyl ketone
Purity: 99%
CAS No.: 765-43-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 16°(61°F)
Refractive Index: 1.4240
Melting Point:
Vzorec:
C₅H₈O MW: 84,12 g/mol Bod varu: 113…114 °C Bod tání: < –70 °C Hustota: 0,898 Bod vzplanutí: 16 °C (61 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001297 CAS číslo: 765-43-5 EINECS: 212-146-4 UN: 1992 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...