Tisk…

1,2-Epoxycyclohexane ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13185.0E A13185.22 A13185.36 AB132975.0100 AB132975.0500
A13185.0EEA 17780 CZK
A13185.0E A13185.22 A13185.36 ABCRAB132975.0100 ABCRAB132975.0500
1,2-Epoxycyclohexane ≥98%
1,2-Epoxycyclohexane

Purity: 98+% CAS No.: 286-20-4 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 24°(75°F) Refractive Index: 1.4520 Melting Point: -40° Boiling Point: 129-130° Solubility: Insoluble in water Beilstein Reference: 383568 UN No.: 2920 MDL No.: MFCD00005162

Vzorec: C₆H₁₀O
Bod varu: 129…130 °C
Bod tání: –40 °C
Hustota: 0,972
Bod vzplanutí: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS číslo: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR