Tisk…

Tetrafluoro-p-benzoquinone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

M-Clarity™ quality level = MQ100

104353-1G 104353-5G
SIAL104353-1GEA 2460 CZK
SIAL104353-1G SIAL104353-5G
Tetrafluoro-p-benzoquinone, Sigma-Aldrich®
Tetrafluoro-p-benzoquinone
Vzorec: C₆F₄O₂
MW: 180,06 g/mol
Bod varu: 133 °C (1013 hPa)
MDL Number: MFCD00001592
CAS číslo: 527-21-9
EINECS: 208-411-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR