ω-Bromo-4-methylacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide
M-Clarity™ quality level = MQ100
159530-100G
159530-25G
159530-5G
SIAL159530-100GEA
8030
CZK
SIAL159530-100G
SIAL159530-25G
SIAL159530-5G
ω-Bromo-4-methylacetophenone, Sigma-Aldrich®
ω-Bromo-4-methylacetophenone
Vzorec:
C₉H₉BrO MW: 213,07 g/mol Bod varu: 105 °C (0,1 mmHg) Bod tání: 95…97 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000203 CAS číslo: 619-41-0 EINECS: 210-595-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...