Tisk…

ω-Bromo-4-methylacetophenone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

B25217.06 B25217.14 B25217.22
B25217.06EA 1700 CZK
B25217.06 B25217.14 B25217.22
ω-Bromo-4-methylacetophenone ≥98%
ω-Bromo-4-methylacetophenone

Purity: 98% CAS No.: 619-41-0 Molecular Formula: C9H9BrO Formula Weight: 213.08 Flash Point: 110°(230°F) Melting Point: 50-54° Boiling Point: 105°/0.1mm Beilstein Reference: 607041 MDL No.: MFCD00000203

Vzorec: C₉H₉BrO
MW: 213.07
Bod varu: 105 °C (0.1 mmHg)
Bod tání: 50…54 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000203
CAS číslo: 619-41-0
EINECS: 210-595-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR