Tisk…

Dibenzoylmethane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D33454-100G D33454-25G
SIALD33454-100GEA 5160 CZK
SIALD33454-100G SIALD33454-25G
Dibenzoylmethane, Sigma-Aldrich®
Dibenzoylmethane
Vzorec: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
Bod varu: 357 °C (1013 hPa)
Bod tání: 74…77 °C
Hustota: 0,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003085
CAS číslo: 120-46-7
EINECS: 204-398-9
Merck Index: 13,03033

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR