Dibenzoylmethane ≥98%
Dodavatel: Thermo Scientific
Synonyms:
1,3-Diphenyl-1,3-propanedione, 1,3-Diphenylpropane-1,3-dione
A13450.14
A13450.22
A13450.36
A13450.14EA
1440
CZK
A13450.14
A13450.22
A13450.36
Dibenzoylmethane ≥98%
Dibenzoylmethane
Purity: 98+%
CAS No.: 120-46-7
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 76-80°
Boiling Point: 219-221°/18mm
Merck Reference: 14,3009
MDL No.: MFCD00003085
Vzorec:
(C₆H₅)C(O)CH₂C(O)(C₆H₅) MW: 224,26 g/mol Bod varu: 219…221 °C (18 mmHg) Bod tání: 76…80 °C Hustota: 0,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003085 CAS číslo: 120-46-7 EINECS: 204-398-9 Merck Index: 14,3009 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...