Chloramin T trihydrát, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
23270-250G
23270-50G
23270-1KG
SIAL23270-250GEA
1390
CZK
SIAL23270-250G
SIAL23270-50G
SIAL23270-1KG
Chloramin T trihydrát, Sigma-Aldrich®
Chloramin T trihydrát
Vzorec:
CH₃C₆H₄SO₂NClNa·3H₂O Bod tání: 167…170 °C Hustota: 1,43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149066 CAS číslo: 7080-50-4 EINECS: 204-854-7 UN: 3263 ADR: 8,III Merck Index: 13,02085 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...