Tisk…

1-Heptyl-4-iodobenzene ≥98%

Dodavatel: Thermo Scientific
Varování

H27017.09 H27017.03
H27017.09EA 4530 CZK
H27017.09 H27017.03
1-Heptyl-4-iodobenzene ≥98%
1-Heptyl-4-iodobenzene

Purity: 98% CAS No.: 131894-91-2 Molecular Formula: C13H19I Formula Weight: 302.19 Flash Point: 120°(248°F) Boiling Point: 165°/10mm MDL No.: MFCD07777084

Vzorec: C₁₃H₁₉I
MW: 302,2 g/mol
Bod varu: 165 °C (10 mmHg)
Bod vzplanutí: >120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD07777084
CAS číslo: 131894-91-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR