2'-Hydroxy-5'-methoxyacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone
114995-25G
114995-5G
SIAL114995-25GEA
7790
CZK
SIAL114995-25G
SIAL114995-5G
2'-Hydroxy-5'-methoxyacetophenone, Sigma-Aldrich®
2'-Hydroxy-5'-methoxyacetophenone
Vzorec:
C₉H₁₀O₃ MW: 166,18 g/mol Bod varu: 281 °C (1013 hPa) Bod tání: 113 °C |
MDL Number:
MFCD00008731 CAS číslo: 705-15-7 EINECS: 211-882-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...