Tisk…

2-Chloro-6-methylphenyl isocyanate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

L11645.03 L11645.06
L11645.03EA 937 CZK
L11645.03 L11645.06
2-Chloro-6-methylphenyl isocyanate ≥97%
2-Chloro-6-methylphenyl isocyanate

Purity: 97% CAS No.: 40398-01-4 Molecular Formula: C8H6ClNO Formula Weight: 167.60 Storage Temperature: 2°C to 8°C Flash Point: 107°(224°F) Refractive Index: 1.5570 Boiling Point: 79-80°/2mm Beilstein Reference: 2690768 UN No.: 2206 MDL No.: MFCD00037030

Vzorec: C₈H₆ClNO
MW: 167,59 g/mol
Bod varu: 79…80 °C (2 mmHg)
Hustota: 1,231
Bod vzplanutí: 107 °C (224 °F)
MDL Number: MFCD00037030
CAS číslo: 40398-01-4
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR