2-Chloro-4'-fluoroacetophenone ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone
B22188.14
B22188.22
B22188.36
B22188.14EA
869
CZK
B22188.14
B22188.22
B22188.36
2-Chloro-4'-fluoroacetophenone ≥99%
2-Chloro-4'-fluoroacetophenone
Purity: 99%
CAS No.: 456-04-2
Molecular Formula: C8H6ClFO
Formula Weight: 172.59
Flash Point:
Vzorec:
C₈H₆ClFO MW: 172,59 g/mol Bod varu: 247 °C Bod tání: 46…50 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011652 CAS číslo: 456-04-2 EINECS: 207-256-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...