Tisk…

ω-Bromo-3-nitroacetophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone

M-Clarity™ quality level = MQ100

344214-25G 344214-5G
SIAL344214-25GEA 2450 CZK
SIAL344214-25G SIAL344214-5G
ω-Bromo-3-nitroacetophenone, Sigma-Aldrich®
ω-Bromo-3-nitroacetophenone
Vzorec: C₈H₆BrNO₃
MW: 244,04 g/mol
Bod tání: 90…94 °C
MDL Number: MFCD00024512
CAS číslo: 2227-64-7
EINECS: 218-764-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR