Tisk…

Benzeneselenenyl chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride

M-Clarity™ quality level = MQ100

183342-10G 183342-1G 183342-50G
SIAL183342-10GEA 5560 CZK
SIAL183342-10G SIAL183342-1G SIAL183342-50G
Benzeneselenenyl chloride, Sigma-Aldrich®
Benzeneselenenyl chloride
Vzorec: (C₆H₅)SeCl
MW: 191.52
Bod varu: 237
Bod tání: 61…62 °C
MDL Number: MFCD00000478
CAS číslo: 5707-04-0
EINECS: 227-196-2
UN: 2928
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR