Benzeneselenenyl chloride 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride
179510100
179510250
179510500
179510010
ACRO179510100EA
4750
CZK
ACRO179510100
ACRO179510250
ACRO179510500
ACRO179510010
Benzeneselenenyl chloride 98%
Benzeneselenenyl chloride
Vzorec:
(C₆H₅)SeCl Bod varu: 120 °C (20 mmHg) Bod tání: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000478 CAS číslo: 5707-04-0 EINECS: 227-196-2 UN: 2811 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | Orange to dark red Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C (Dec) |
Titration Argentometric | ≥97.5 % |
Crosslinking | 25 % DVB |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...