Fenol, crystals, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
M-Clarity™ quality level = MQ200
242322-1.5KG
242322-25G
242322-500G
SIAL242322-1.5KGEA
6810
CZK
SIAL242322-1.5KG
SIAL242322-25G
SIAL242322-500G
Fenol, crystals, Sigma-Aldrich®
Fenol
Vzorec:
C₆H₅OH MW: 94.11 Bod varu: 181.7 Bod tání: 40.8 Hustota: 1.06 g/cm³ (20 °C) Bod vzplanutí: 81 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS číslo: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1 II Merck Index: 12,07390 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...