Fenol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
M-Clarity™ quality level = MQ200
P5566-100G
P5566-25G
P5566-500G
SIALP5566-100GEA
2700
CZK
SIALP5566-100G
SIALP5566-25G
SIALP5566-500G
Fenol, Sigma-Aldrich®
Fenol
Vzorec:
C₆H₅OH Bod varu: 181,7 °C (1013 hPa) Hustota: 1,0722 g/cm³ (20 °C) Bod vzplanutí: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS číslo: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...