4-Fluoro-2-methoxyphenol ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B22211.06
B22211.14
B22211.06EA
4330
CZK
B22211.06
B22211.14
4-Fluoro-2-methoxyphenol ≥97%
4-Fluoro-2-methoxyphenol
Purity: 97%
CAS No.: 450-93-1
Molecular Formula: C7H7FO2
Formula Weight: 142.13
Flash Point: 102°(216°F)
Refractive Index: 1.5170
Odour: Phenol-like
Boiling Point: 195°
Beilstein Reference: 5425200
MDL No.: MFCD00070797
Vzorec:
C₇H₇FO₂ MW: 142,13 g/mol Bod varu: 195 °C Hustota: 1,25 Bod vzplanutí: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070797 CAS číslo: 450-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...