2-Fluoro-6-methoxyphenol ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H26471.03
H26471.06
H26471.03EA
2630
CZK
H26471.03
H26471.06
2-Fluoro-6-methoxyphenol ≥97%
2-Fluoro-6-methoxyphenol
Purity: 97%
CAS No.: 73943-41-6
Molecular Formula: C7H7FO2
Formula Weight: 142.13
Flash Point: 103°(218°F)
Refractive Index: 1.52
Boiling Point: 130-131°/36mm
MDL No.: MFCD00075140
Vzorec:
C₇H₇FO₂ MW: 142,13 g/mol Bod varu: 130…131 °C (36 mmHg) Hustota: 1,23 Bod vzplanutí: 103 °C (218 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075140 CAS číslo: 73943-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...