Tisk…

5-Hydroxysophthalic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Hydroxyisophthalic acid

A13845.0B A13845.18 A13845.30
A13845.0BEA 12390 CZK
A13845.0B A13845.18 A13845.30
5-Hydroxysophthalic acid ≥98%
5-Hydroxysophthalic acid

Purity: 98% CAS No.: 618-83-7 Molecular Formula: C8H6O5 Formula Weight: 182.13 Odour: Odourless Melting Point: ca 300° dec. Beilstein Reference: 2693561 MDL No.: MFCD00002515

Vzorec: C₈H₆O₅
MW: 182,13 g/mol
Bod varu: 386 °C (1013 hPa)
Bod tání: ∼ 300 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00002515
CAS číslo: 618-83-7
EINECS: 210-565-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR