Tisk…

5-Aminoisophthalic acid 95%

Dodavatel: Thermo Scientific
Nebezpečí

A15074.18 A15074.30
A15074.18EA 1270 CZK
A15074.18 A15074.30
5-Aminoisophthalic acid 95%
5-Aminoisophthalic acid

Purity: 95% CAS No.: 99-31-0 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 300° Beilstein Reference: 2805628

Vzorec: C₈H₇NO₄
MW: 181,15 g/mol
Bod varu: 515 °C (1013 hPa)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002522
CAS číslo: 99-31-0
EINECS: 202-748-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR