Tisk…

Dimethyl-5-hydroxyisophthalate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Hydroxyisophthalic acid dimethyl ester, Dimethyl 5-hydroxyisophthalate

M-Clarity™ quality level = MQ100

371785-10G 371785-50G
SIAL371785-10GEA 1580 CZK
SIAL371785-10G SIAL371785-50G
Dimethyl-5-hydroxyisophthalate, Sigma-Aldrich®
Dimethyl-5-hydroxyisophthalate
Vzorec: C₁₀H₁₀O₅
MW: 210,19 g/mol
MDL Number: MFCD00134367
CAS číslo: 13036-02-7
EINECS: 235-899-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR