Tisk…

Dimethyl-5-nitroisophthalate ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dimethyl 5-nitroisophthalate, 5-Nitroisophthalic acid dimethyl ester

A10610.0B A10610.30
A10610.0BEA 7310 CZK
A10610.0B A10610.30
Dimethyl-5-nitroisophthalate ≥98%
Dimethyl-5-nitroisophthalate

Purity: 98+% CAS No.: 13290-96-5 Molecular Formula: C10H9NO6 Formula Weight: 239.19 Melting Point: 121-123° Beilstein Reference: 2140916

Vzorec: C₁₀H₉NO₆
MW: 239.18
Bod tání: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00008429
CAS číslo: 13290-96-5
EINECS: 236-307-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR