Tisk…

Dimethyl-5-aminoisophthalate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dimethyl 5-aminoisophthalate, 5-Aminoisophthalic acid dimethyl ester

M-Clarity™ quality level = MQ100

D142158-100G D142158-25G
SIALD142158-100GEA 2920 CZK
SIALD142158-100G SIALD142158-25G
Dimethyl-5-aminoisophthalate, Sigma-Aldrich®
Dimethyl-5-aminoisophthalate
Vzorec: C₁₀H₁₁NO₄
MW: 209,2 g/mol
MDL Number: MFCD00008435
CAS číslo: 99-27-4
EINECS: 202-743-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR