Dimethyl-5-aminoisophthalate 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dimethyl 5-aminoisophthalate, 5-Aminoisophthalic acid dimethyl ester
B20761.14
B20761.22
B20761.14EA
1080
CZK
B20761.14
B20761.22
Dimethyl-5-aminoisophthalate 98%
Dimethyl-5-aminoisophthalate
Purity: 98%
CAS No.: 99-27-4
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Melting Point: 178-182°
Beilstein Reference: 2116565
Vzorec:
C₁₀H₁₁NO₄ MW: 209,2 g/mol Bod tání: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008435 CAS číslo: 99-27-4 EINECS: 202-743-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...