Tisk…

Methyl-3,4,5-trihydroxybenzoate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L05926.18 L05926.30 L05926.0250
L05926.18EA 1700 CZK
L05926.18 L05926.30 ABCRL05926.0250
Methyl-3,4,5-trihydroxybenzoate ≥98%
Methyl-3,4,5-trihydroxybenzoate

Purity: 98% CAS No.: 99-24-1 Molecular Formula: C8H8O5 Formula Weight: 184.15 Melting Point: 198-203° Beilstein Reference: 2113180 Merck Reference: 14,4345 MDL No.: MFCD00002194

Vzorec: C₈H₈O₅
Bod varu: 450 °C (1013 hPa)
Bod tání: 198…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00002194
CAS číslo: 99-24-1
EINECS: 202-741-7
Merck Index: 14,4345

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR