Tisk…

Isophthaloyl dichloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Isophthaloyl chloride

M-Clarity™ quality level = MQ200

I19403-100G I19403-500G
SIALI19403-100GEA 1920 CZK
SIALI19403-100G SIALI19403-500G
Isophthaloyl dichloride, Sigma-Aldrich®
Isophthaloyl dichloride
Vzorec: C₈H₄Cl₂O₂
MW: 203,02 g/mol
Bod varu: 276 °C (1013 hPa)
Bod tání: 42…45 °C
Hustota: 1,388 g/cm³ (17 °C)
MDL Number: MFCD00000678
CAS číslo: 99-63-8
EINECS: 202-774-7
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR