Tisk…

4-Chlor-2-fluorfenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

334227-25G 334227-5G
SIAL334227-25GEA 0 CZK
SIAL334227-25G SIAL334227-5G
4-Chlor-2-fluorfenol, Sigma-Aldrich®
4-Chlor-2-fluorfenol
Vzorec: C₆H₄ClFO
MW: 146,55 g/mol
MDL Number: MFCD00075030
CAS číslo: 348-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR