Tisk…

4-Bromo-2-methoxyphenol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L08801.09 L08801.18 L08801.04
L08801.09EA 4090 CZK
L08801.09 L08801.18 L08801.04
4-Bromo-2-methoxyphenol ≥98%
4-Bromo-2-methoxyphenol

Purity: 98% CAS No.: 7368-78-7 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Odour: Phenol-like Melting Point: 32-37° Boiling Point: 129-132°/12mm Beilstein Reference: 2045286 MDL No.: MFCD00051937

Vzorec: C₇H₇BrO₂
Bod varu: 129…132 °C (12 mmHg)
Bod tání: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00051937
CAS číslo: 7368-78-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR