Tisk…

2-Bromo-6-methoxyphenol ≥98%

Dodavatel: Thermo Scientific
Varování

B25535.03 B25535.06 B25535.14
B25535.03EA 1500 CZK
B25535.03 B25535.06 B25535.14
2-Bromo-6-methoxyphenol ≥98%
2-Bromo-6-methoxyphenol

Purity: 98+% CAS No.: 28165-49-3 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Melting Point: 62-65° MDL No.: MFCD08146628

Vzorec: C₇H₇BrO₂
MW: 203,04 g/mol
Bod tání: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08146628
CAS číslo: 28165-49-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR