4-Bromoisophthalic acid 96%
Dodavatel: Thermo Scientific
|
Varování
|
A13152.06
A13152.14
A13152.22
A13152.06EA
2510
CZK
A13152.06
A13152.14
A13152.22
4-Bromoisophthalic acid 96%
4-Bromoisophthalic acid
Purity: 96%
CAS No.: 6939-93-1
Molecular Formula: C8H5BrO4
Formula Weight: 245.03
Melting Point: 296-302°
Beilstein Reference: 1959392
Vzorec:
C₈H₅BrO₄ MW: 245,03 g/mol Bod tání: 296…302 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002404 CAS číslo: 6939-93-1 EINECS: 230-078-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...