Tisk…

2-Bromo-6-chloro-4-fluoroaniline

Dodavatel: Merck


M-Clarity™ quality level = MQ100

569283-1G 569283-5G
SIAL569283-1GEA 826 CZK
SIAL569283-1G SIAL569283-5G
2-Bromo-6-chloro-4-fluoroaniline
2-Bromo-6-chloro-4-fluoroaniline
Vzorec: C₆H₄BrClFN
MW: 224,46 g/mol
Bod varu: 260 °C (1013 hPa)
Bod tání: 55…56 °C
MDL Number: MFCD00051707
CAS číslo: 201849-14-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR