Penicillin G sodium salt, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
M-Clarity™ quality level = MQ200
P3032-1MU
P3032-100MU
P3032-10MU
P3032-25MU
SIALP3032-1MUEA
832
CZK
SIALP3032-1MU
SIALP3032-100MU
SIALP3032-10MU
SIALP3032-25MU
Penicillin G sodium salt, Sigma-Aldrich®
Penicillin G sodium salt
Vzorec:
C₁₆H₁₇N₂NaO₄S MW: 356,38 g/mol Bod tání: 255 °C Hustota: 1,38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00069666 CAS číslo: 69-57-8 EINECS: 200-710-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...