Tisk…

MOPS-Na, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt

M-Clarity™ quality level = MQ400

M5789-100G M5789-1KG M5789-250G M5789-25G
SIALM5789-100GEA 17520 CZK
SIALM5789-100G SIALM5789-1KG SIALM5789-250G SIALM5789-25G
MOPS-Na, Sigma-Aldrich®
MOPS-Na
Vzorec: C₇H₁₄NNaO₄S
MW: 231,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064350
CAS číslo: 71119-22-7
EINECS: 428-420-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR