Tisk…

MES sodium salt, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: MES-Na

M-Clarity™ quality level = MQ400

M3885-100G M3885-25G
SIALM3885-100GEA 3910 CZK
SIALM3885-100G SIALM3885-25G
MES sodium salt, Sigma-Aldrich®
MES sodium salt
Vzorec: C₆H₁₂NNaO₄S
MW: 217.22
MDL Number: MFCD00065473
CAS číslo: 71119-23-8
EINECS: 275-203-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR