Tisk…

MOPS, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

M-Clarity™ quality level = MQ300

M3183-100G M3183-1KG M3183-25G M3183-500G M3183-6X100G M3183-6X500G
SIALM3183-100GEA 5280 CZK
SIALM3183-100G SIALM3183-1KG SIALM3183-25G SIALM3183-500G SIALM3183-6X100G SIALM3183-6X500G
MOPS, Sigma-Aldrich®
MOPS
Vzorec: C₇H₁₅NO₄S
MW: 209,26
Bod tání: > 250 °C
Bod vzplanutí: 110 °C (ot. pohárek)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS číslo: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR