Tisk…

MES, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid

M-Clarity™ quality level = MQ400

M3671-1KG M3671-250G M3671-50G
SIALM3671-1KGEA 0 CZK
SIALM3671-1KG SIALM3671-250G SIALM3671-50G
MES, Sigma-Aldrich®
MES
Vzorec: C₆H₁₃NO₄S
MW: 195,24 g/mol
Bod tání: 316 °C
Storage Temperature: Ambient
MDL Number: MFCD00006181
CAS číslo: 4432-31-9
EINECS: 224-632-3
Merck Index: 13,05929

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR